missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APP Antibody (Hu15) [DyLight 488], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals FAB9710K
This item is not returnable.
View return policy
Description
APP Monoclonal antibody specifically detects APP in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| APP | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| Hu15 | |
| Flow Cytometry, CyTOF-ready | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| Human APP/Protease Nexin II | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 351 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction