Learn More
APP Mouse anti-Human, Clone: OTI4C9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505011
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa.Specifications
| APP | |
| Monoclonal | |
| Unconjugated | |
| APP | |
| AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2, preA4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 351 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P05067 | |
| APP | |
| Full length human recombit protein of human APP produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.