missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL Antibody (OTI4B11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71782AF350
This item is not returnable.
View return policy
Description
APPL Monoclonal specifically detects APPL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APPL | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| APPL1 | |
| Full length recombinant protein of human APPL (NP_036228) produced in HEK293T cells. | |
| 0.1 mL | |
| 26060.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4B11 | |
| Western Blot, Immunohistochemistry | |
| Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction