Learn More
APPL1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12639-100UL
 View more versions of this product
Product Code. 18016685
  • € 450.00 / 100µL
Description
The APPL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human APPL1. This antibody recognizes human, mouse, rat antigen. The APPL1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
APPL1
1000 μg/mL
APPL, APPL1, Dip13 alpha, DIP13A, DIP13alpha, KIAA1428
100 μL
26060
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa