missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL2 Monoclonal Antibody (OTI8F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812391
This item is not returnable.
View return policy
Description
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.Specifications
| APPL2 | |
| Monoclonal | |
| Unconjugated | |
| APPL2 | |
| DIP13B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55198 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8F1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NEU8 | |
| APPL2 | |
| Full length human recombit protein of human APPL2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction