missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568745
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%; Pig: 85%; Rabbit: 93%; Rat: 79%.
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
Specifications
| APR3 | |
| Polyclonal | |
| Unconjugated | |
| ATRAID | |
| 0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; all-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein APR-3; apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; C11H2orf28; C17H2orf28; C2orf28; H1E6 protein; HSPC013; LOW QUALITY PROTEIN: all-trans retinoic acid-induced differentiation factor; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UW56 | |
| ATRAID | |
| synthetic peptide directed towards the middle region of human APR3 (aa 127-176). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction