Learn More
Aprepitant, Mikromol™
Description
Aprepitant
Specifications
Specifications
| Chemical Name or Material | Aprepitant |
| CAS | 170729-80-3 |
| Analyte or Component Names | Aprepitant |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H21 F7 N4 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 5-[[(2R,3S)-2-[(1R)-1-[3,5-bis(Trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one, Aprepitant, Emend, L 754030, MK 0869, MK 869, ONO 7436, 3H-1,2,4-Triazol-3-one, 5-[[2-[1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-, [2R-[2alpha(R*),3alpha]]- |
| InChI Formula | InChI=1S/C23H21F7N4O3/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)37-20-19(13-2-4-17(24)5-3-13)34(6-7-36-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19+,20-/m1/s1 |
| SMILES | C[C@@H](O[C@H]1OCCN(CC2=NNC(=O)N2)[C@H]1c3ccc(F)cc3)c4cc(cc(c4)C(F)(F)F)C(F)(F)F |
| IUPAC Name | 3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-1,4-dihydro-1,2,4-triazol-5-one |
| Show More |