missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APRIL/TNFSF13 Antibody (670840) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8843I
This item is not returnable.
View return policy
Description
APRIL/TNFSF13 Monoclonal antibody specifically detects APRIL/TNFSF13 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| APRIL/TNFSF13 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 670840 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
| Human embryonic kidney cell line HEK293EBNA-derived recombinant human APRIL/TNFSF13, Ala105-Leu250, Accession # O75888 | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 8741 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction