Learn More
APRIN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-90122
 View more versions of this product
Product Code. 18779653
  • € 624.00 / 0.10mL
Description
APRIN Polyclonal specifically detects APRIN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence, Immunohistochemistry (Paraffin)
Rabbit
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
0.1 mL
Mouse, Rat