missing translation for 'onlineSavingsMsg'
Learn More

Aprocitentan, MedChemExpress

Product Code. 16466908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466908 10 mg 10mg
16499437 5 mg 5mg
16462967 50 mg 50mg
16436986 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466908 Supplier Medchem Express Supplier No. HY15895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively.

Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively.

Specifications

Chemical Name or Material Aprocitentan
CAS 1103522-45-7
Color White
Purity Grade Notes Research
Molecular Formula C16H14Br2N6O4S
Quantity 10 mg
Synonym ACT-132577
Solubility Information DMSO : ≥ 46 mg/mL (84.22 mM)
SMILES O=S(N)(NC1=NC=NC(OCCOC2=NC=C(Br)C=N2)=C1C3=CC=C(Br)C=C3)=O
Molecular Weight (g/mol) 546.19
Formula Weight 546.19
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.