Learn More
APS-2-79, MedChemExpress
Product Code. 16495837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495837 5 mg 5mg
16453108 10 mg 10mg
16442626 25 mg 25mg
16419666 50 mg 50mg
16414406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16495837 Supplier Medchem Express Supplier No. HY100627
MedChemExpress APS-2-79 is a KSR-dependent MEK antagonist. APS-2-79 inhibits ATPbiotin binding to KSR2 within the KSR2-MEK1 complexe with an IC50 of 120 nM. APS-2-79 makes the stabilization of the KSR inactive state antagonizes oncogenic Ras-MAPK signaling.
APS-2-79 is a KSR-dependent MEK antagonist. APS-2-79 inhibits ATPbiotin binding to KSR2 within the KSR2-MEK1 complexe with an IC50 of 120 nM. APS-2-79 makes the stabilization of the KSR inactive state antagonizes oncogenic Ras-MAPK signaling.
Specifications
Chemical Name or Material APS-2-79
CAS 2002381-25-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H21N3O3
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (51.62 mM; Need ultrasonic)
SMILES CC1=CC(OC2=CC=CC=C2)=CC=C1NC3=NC=NC4=CC(OC)=C(OC)C=C43
Molecular Weight (g/mol) 387.43
Formula Weight 387.43
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only