Learn More
AQP1 Mouse anti-Human, Clone: OTI2D10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502357
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.Specifications
| AQP1 | |
| Monoclonal | |
| Unconjugated | |
| AQP1 | |
| AQP-CHIP, CHIP28, CO | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 358 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P29972 | |
| AQP1 | |
| Full length human recombit protein of human AQP1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.