Learn More
AQP8 Mouse anti-Human, Clone: OTI6E6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810725
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.Specifications
| AQP8 | |
| Monoclonal | |
| Unconjugated | |
| AQP8 | |
| AQP-8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 343 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6E6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O94778 | |
| AQP8 | |
| Full length human recombit protein of human AQP8 produced in HEK293T. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.