Learn More
Buerkle™ Aqua Sampler
Product Code. 10188701 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
350 mL
700 mL
1050 mL
Length (Metric):
40 cm
76 cm
107 cm
Unit Size:
Each
Product Code. 10188701
Brand: Buerkle™ 53081050
Only null left
Add to basket
Add to basket
Sampler for ground water samples from wells, pits and other sources.
  • Fits all normal well shafts with control openings to 2in diameter, without tilting or getting stuck
  • Ultra-pure materials guarantee the highest degree of sample purity
  • Easy to clean thanks to rounded edges and threads
  • FEP/PTFE ultra-pure, anti-adhesive surface
  • Without cable
TRUSTED_SUSTAINABILITY
Specifications
Product Type AquaSampler
Material PTFE, FEP
Capacity (Metric) 1050 mL
For Use With (Application) For Ground Water Samples From Wells, Pits and Other Sources
Diameter (Metric) Outer 46 mm
Length (Metric) 107 cm