Learn More
3M™ Nomad™ Aqua Textile Drop Down Mat, Series 85
Product Code. 17188570
Change view
Click to view available options
Dimensions (L x W):
150 x 91.4 cm
200 x 130 cm
300 x 130 cm
300 x 200 cm
600 x 130 cm
Color:
Black
Gray
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color unitSize
17188570 150 x 91.4 cm Gray Each
17128580 150 x 91.4 cm Black Each
17178590 200 x 130 cm Black Each
17198570 200 x 130 cm Gray Each
17108630 300 x 130 cm Gray Each
17108580 300 x 200 cm Gray Each
17138580 600 x 130 cm Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17188570 Supplier 3M™ Nomad™ Supplier No. 7000032560
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Advanced water removal and retention properties keep floors cleaner and safer, for an extended period of time. Traditional, deep cut pile and coir matting allows dirt and moisture to sit on the surface and be tracked into the building.
  • High performing textile entrance mat, with advanced water removal and retention properties
  • Stops 80% of dirt and moisture at the door
  • Entrance mat for high traffic applications (1500 to 5000 crossings per day)
  • Phthalate-free, slip-resistant PVC backing
  • Excellent appearance retention
  • Long lasting and easy to clean
  • Available as drop down mats or in rolls
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 150 x 91.4 cm
For Use With (Application) Heavy traffic areas
Model Nomad™ Aqua, Series 85
Resistance Slip-resistant
Thickness (Metric) 7.5 mm
Color Gray
Length (Metric) 150 cm
Material PVC (Backing)
Width (Metric) 91.4 cm