Learn More
Invitrogen™ Aquaporin 4 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA578812
Product Code. 15944395
  • € 452.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, rat brain tissue, mouse brain tissue, mouse brain tissue. IHC: human lung cancer tissue, mouse brain tissue, rat brain tissue IHC-F: Mouse Brain tissue, Rat Brain tissue.
In skeletal muscle, AQP4 (aquaporin 4 also known as mercurial insensitive water channel), localizes to the sarcolemma of fast-twitch muscle fibers. Aquaporins (AQPs) are a large family of integral membrane water transport channel proteins that facilitate the transport of water through the cell membrane. This function is conserved in animals, plants and bacteria. Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin 4
Polyclonal
Unconjugated
AQP4
Aqp4; AQP-4; aquaporin 4; aquaporin type4; Aquaporin4; aquaporin-4; HMIWC2; Mercurial-insensitive water channel; MGC22454; MIWC; MIWC2; WCH4
Rabbit
Antigen affinity chromatography
RUO
11829, 25293, 361
-20°C
Lyophilized
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 4mg trehalose and no preservative
P47863, P55087, P55088
AQP4
A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 4 (295-323aa KPGVVHVIDVDRGEEKKGKDQSGEVLSSV).