missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-67247MFV610
This item is not returnable.
View return policy
Description
Aquaporin-5 Polyclonal antibody specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Aquaporin-5 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| AQP-5, aquaporin 5, aquaporin-5 | |
| Synthetic peptide made to an N-terminal portion of the human Aquaporin-5 protein (between amino acids 170-250). [UniProt P55064] | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 362 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction