missing translation for 'onlineSavingsMsg'
Learn More

Aquavan, TRC

Product Code. 30338293
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338293 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338293 Supplier Toronto Research Chemicals Supplier No. TRCA73544510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aquavan
CAS 258516-87-9
Molecular Formula C13H19Na2O5P
Quantity 10 mg
Synonym 1-[2,6-bis(1-methylethyl)phenoxy]methanol 1-(Dihydrogen phosphate) Disodium Salt, [2,6-Bis(1-methylethyl)phenoxy]methanol Dihydrogen Phosphate Disodium Salt, Aqavan Injection, Fospropofol Disodium, GPI 15715, Lusedra
InChI Formula InChI=1S/C13H21O5P.2Na/c1-9(2)11-6-5-7-12(10(3)4)13(11)17-8-18-19(14,15)16;;/h5-7,9-10H,8H2,1-4H3,(H2,14,15,16);;/q;2*+1/p-2
SMILES [Na+].[Na+].CC(C)c1cccc(C(C)C)c1OCOP(=O)([O-])[O-]
IUPAC Name disodium;[2,6-di(propan-2-yl)phenoxy]methyl phosphate
Molecular Weight (g/mol) 332.24
Formula Weight 332.0765
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.