Learn More
(aR)-5,6,7,8-Tetrahydro-a-methyl-1-naphthalenemethanamine, TRC
Specifications
Specifications
| Chemical Name or Material | (alphaR)-5,6,7,8-Tetrahydro-alpha-methyl-1-naphthalenemethanamine |
| CAS | 851984-49-1 |
| Molecular Formula | C12H17N |
| Quantity | 250 mg |
| Synonym | (αR)-5,6,7,8-Tetrahydro-α-methyl-1-naphthalenemethanamine (ACI), (1R)-1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethan-1-amine, (1R)-1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethanamine, 1-Naphthalenemethanamine 5,6,7,8-tetrahydro-α-methyl-','(αR) |
| InChI Formula | InChI=1S/C12H17N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h4,6,8-9H,2-3,5,7,13H2,1H3/t9-/m1/s1 |
| SMILES | C[C@H](C1=C2C(CCCC2)=CC=C1)N |
| IUPAC Name | (R)-1-(5,6,7,8-tetrahydronaphthalen-1-yl)ethan-1-amine |
| Molecular Weight (g/mol) | 175.27 |
| Formula Weight | 175.14 |