Learn More
(aR)-a-Methyl-N-[3-(3-methylphenyl)propyl]-1-naphthalenemethanamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (alphaR)-alpha-Methyl-N-[3-(3-methylphenyl)propyl]-1-naphthalenemethanamine Hydrochloride |
| CAS | 253337-60-9 |
| Molecular Formula | C22 H25 N |
| Quantity | 500 mg |
| Synonym | (αR)-α-Methyl-N-[3-(3-methylphenyl)propyl]-1-naphthalenemethanamine |
| InChI Formula | InChI=1S/C22H25N/c1-17-8-5-9-19(16-17)10-7-15-23-18(2)21-14-6-12-20-11-3-4-13-22(20)21/h3-6,8-9,11-14,16,18,23H,7,10,15H2,1-2H3/t18-/m1/s1 |
| SMILES | C[C@@H](NCCCc1cccc(C)c1)c2cccc3ccccc23 |
| IUPAC Name | 3-(3-methylphenyl)-N-[(1R)-1-naphthalen-1-ylethyl]propan-1-amine |
| Molecular Weight (g/mol) | 303.44 |
| Formula Weight | 303.1987 |