Learn More
Hirschmann™ Class AS Graduated Pipettes
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes are available in either blue or amber graduation.
€ 82.20 - € 159.00
Specifications
Class AS
Material AR-GLAS
Graduated Yes
Type Graduated Pipet
View More Specs
Products 18
Product Code Brand Volume (Metric) Graduations Graduation Color Tolerance Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Graduation Color Tolerance Price Quantity & Availability  
11682289
View accessory products View Documents Promotion Details
Hirschmann™
1100133
1 mL 0.01 mL Amber 0.007 mL
€ 82.20
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11767951
View accessory products View Documents Promotion Details
Hirschmann™
1100160
10 mL 0.1 mL Amber 0.05 mL
€ 95.40
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
11720814
View accessory products View Documents Promotion Details
Hirschmann™
1101129
0.5 mL 0.01 mL Blue 0.006 mL
€ 105.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11786612
View accessory products View Documents Promotion Details
Hirschmann™
1100129
0.5 mL 0.01 mL Amber 0.006 mL
€ 105.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11740814
View accessory products View Documents Promotion Details
Hirschmann™
1101135
1 mL 0.1 mL Blue 0.007 mL
€ 83.80
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
11716622
View Documents Promotion Details
Hirschmann™
1100135
1 mL 0.1 mL Amber 0.007 mL
€ 82.20
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11710834
View accessory products View Documents Promotion Details
Hirschmann™
1101165
20 mL 0.1 mL Blue 0.1 mL
€ 94.20
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11737971
View accessory products View Documents Promotion Details
Hirschmann™
1100165
20 mL 0.1 mL Amber 0.1 mL
€ 94.20
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
13114969
View Documents Promotion Details
Hirschmann™
1101140
2 mL 0.1 mL Blue 0.1 mL
€ 108.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
15241209
View Documents Promotion Details
Hirschmann™
1101142
2 mL 0.1 mL Blue 0.01 mL
€ 83.80
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11796612
View accessory products View Documents Promotion Details
Hirschmann™
1100142
2 mL 0.1 mL Amber 0.01 mL
€ 82.20
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11692289
View accessory products View Documents Promotion Details
Hirschmann™
1100141
2 mL 0.02 mL Amber 0.01 mL
€ 82.20
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11762352
View Documents Promotion Details
Hirschmann™
1100175
50 mL 0.2 mL Amber 0.2 mL
€ 159.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11757951
View accessory products View Documents Promotion Details
Hirschmann™
1100153
5 mL 0.05 mL Amber 0.03 mL
€ 96.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11790824
View accessory products View Documents Promotion Details
Hirschmann™
1101154
5 mL 0.1 mL Blue 0.03 mL
€ 90.60
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
11706622
View accessory products View Documents Promotion Details
Hirschmann™
1100154
5 mL 0.1 mL Amber 0.03 mL
€ 90.60
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
11747971
View accessory products View Documents Promotion Details
Hirschmann™
1100170
25 mL 0.1 mL Amber 0.1 mL
€ 94.20
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
11722303
View Documents Promotion Details
Hirschmann™
1100140
2 mL 0.01 mL Amber 0.01 mL
€ 108.00
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN EN ISO 835
  • Main point ring graduation (in either blue or amber)
  • Graduated to tip, zero on tip, type 3
  • With cotton plug end as standard
  • Conformity certified, with dated batch identification
Specifications
Specifications
AS
AR-GLAS
Yes
Graduated Pipet