missing translation for 'onlineSavingsMsg'
Learn More

AR-M 1000390 hydrochloride, MedChemExpress

Product Code. 16489786
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489786

Brand: Medchem Express HY101039A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AR-M 1000390 hydrochloride is an exceptionally selective, potent δ opioid receptor agonist with an EC50 of 7.2±0.9 nM for δ agonist potency.

AR-M 1000390 hydrochloride is an exceptionally selective, potent δ opioid receptor agonist with an EC50 of 7.2±0.9 nM for δ agonist potency.

Specifications

Chemical Name or Material AR-M 1000390 hydrochloride
Physical Form Solid
CAS 209808-47-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H29ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 150 mg/mL (389.67 mM) ∣H2O : 50 mg/mL (129.89 mM; Need ultrasonic)
SMILES O=C(N(CC)CC)C1=CC=C(/C(C2=CC=CC=C2)=C3CCNCC/3)C=C1.Cl
Molecular Weight (g/mol) 384.94
Formula Weight 384.94
Percent Purity 98.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.