missing translation for 'onlineSavingsMsg'
Learn More

AR Mouse anti-Human, Clone: AR441 + DHTR/882, Abnova™

Product Code. 16043136
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16043136 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16043136 Supplier Abnova Supplier No. MAB14823.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human AR.

  • High specificity
  • High reactivity

Specifications

Antigen AR
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone AR441 + DHTR/882
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human AR.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 1 mM PBS.
Gene Accession No. P10275
Gene Alias AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM
Gene Symbols AR
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 302-318 of human AR (AR441) and recombinant protein corresponding to full length human AR (DHTR/882).
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.