missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR Mouse anti-Human, Clone: DHTR/882, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14304.100ug
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Mouse monoclonal antibody raised against full length recombinant human AR.
- High specificity
- High reactivity
Specifikationer
| AR | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P10275 | |
| AR | |
| Recombinant protein corresponding to full length human AR. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| DHTR/882 | |
| Mouse monoclonal antibody raised against full length recombinant human AR. | |
| In 1 mM PBS. | |
| AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 367 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only (RUO)
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering