missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arabidopsis thaliana ACS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117718
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
1-aminocyclopropane-1-carboxylate synthase (ACS) enzymes catalyze the conversion of S-adenosyl-L-methionine (SAM) into 1-aminocyclopropane-1-carboxylate (ACC), a direct precursor of ethylene.
Specifications
| Arabidopsis thaliana ACS8 | |
| Polyclonal | |
| Unconjugated | |
| ACS8 | |
| 1-aminocyclopropane-1-carboxylate synthase - like protein; 1-amino-cyclopropane-1-carboxylate synthase 8; 1-aminocyclopropane-1-carboxylate synthase 8; 1-aminocyclopropane-1-carboxylate synthase ACS8; ACC synthase 8; ACS8; At4g37770; S-adenosyl-L-methionine methylthioadenosine-lyase 8; T28I19.50; T28I19_50 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 829933 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 4.28 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9T065 | |
| ACS8 | |
| Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469aa). | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction