missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Arabidopsis thaliana OSM34 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17280084 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17280084 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17280084 Supplier Invitrogen™ Supplier No. PA5117589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

defense response to bacterium, incompatible interaction, defense response to fungus, incompatible interaction, response to salt stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arabidopsis thaliana OSM34
Applications Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OSM34
Gene Accession No. P50700
Gene Alias AT4G11650; ATOSM34; OSM34; osmotin 34; Osmotin-like protein OSM34; T5C23.80; T5C23_80
Gene Symbols OSM34
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana Osmotin-like protein OSM34 protein (23-244aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 826770
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.