Learn More
ARAF Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803658S
Encompass_Preferred
 View more versions of this product
Product Code. 17522873
  • € 242.00 / 30µL
Description
ARAF Monoclonal antibody specifically detects ARAF in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
ARAF
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P33897
Mouse
Affinity chromatography
RUO
215
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2F8
Unconjugated
ABCD1
1200013E08Rik; Araf; A-RAF; A-Raf proto-oncogene serine/threonine-protein kinase; A-Raf proto-oncogene, serine/threonine kinase; Araf1; A-raf-1; AW495444; Oncogene ARAF1; PKS; PKS2; Proto-oncogene A-Raf; proto-oncogene A-Raf-1; Proto-oncogene Pks; raf a; Raf related protein; RAFA1; raf-related oncogene; Ras-binding protein; Ras-binding protein DA-Raf; RP1-230G1.1; serine/threonine-protein kinase A-Raf; v-raf murine sarcoma 3611 viral oncogene homolog; v-raf murine sarcoma 3611 viral oncogene homolog 1; v-raf murine sarcoma 3611 viral oncogene-like protein; v-raf oncogene homolog 1 (murine sarcoma 3611 virus)
Human recombinant protein fragment corresponding to amino acids 508-745 of human ABCD1 produced in E.coli.
30 μL