missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAF Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803662S
This item is not returnable.
View return policy
Description
ARAF Monoclonal antibody specifically detects ARAF in Human samples. It is validated for Western BlotSpecifications
| ARAF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17405 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6609 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| Smpd1 | |
| 1200013E08Rik; Araf; A-RAF; A-Raf proto-oncogene serine/threonine-protein kinase; A-Raf proto-oncogene, serine/threonine kinase; Araf1; A-raf-1; AW495444; Oncogene ARAF1; PKS; PKS2; Proto-oncogene A-Raf; proto-oncogene A-Raf-1; Proto-oncogene Pks; raf a; Raf related protein; RAFA1; raf-related oncogene; Ras-binding protein; Ras-binding protein DA-Raf; RP1-230G1.1; serine/threonine-protein kinase A-Raf; v-raf murine sarcoma 3611 viral oncogene homolog; v-raf murine sarcoma 3611 viral oncogene homolog 1; v-raf murine sarcoma 3611 viral oncogene-like protein; v-raf oncogene homolog 1 (murine sarcoma 3611 virus) | |
| Human recombinant protein fragment corresponding to amino acids 62-341 of human SMPD1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction