Learn More
ARAF Mouse anti-Human, Clone: OTI2F5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803675
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Several serine/threonine protein kinases have been implicated as intermediates in signal transduction pathways. These include ERK/MAP kinases, ribosomal S6 kinase (Rsk) and Raf-1. Raf-1 is a cytoplasmic protein with intrinsic serine/threonine activity. It is broadly expressed in nearly all cell lines tested to date and is the cellular homolog of v-Raf, the product of the transforming gene of the 3611 strain of murine sarcoma virus. The unregulated kinase activity of the v-Raf protein has been associated with transformation and mitogenesis while the activity of Raf-1 is normally suppressed by a regulatory N-terminal domain. A-Raf, a second member of the Raf gene family of serine/ threonine protein kinases, exhibits substantial homology to Raf-1 within the kinase domain of the two molecules, but less homology elsewhere. Expression of A-Raf is found at highest levels in urogenital tissues and kidney and at lowest level in brain tissue.Specifications
| ARAF | |
| Monoclonal | |
| Unconjugated | |
| ARAF | |
| A-RAF, ARAF1, PKS2, RAFA1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 369 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2F5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P10398 | |
| ARAF | |
| Human recombit protein fragment corresponding to amino acids 145-365 of human ARAF produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.