missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAP1 Antibody (ARAP1-2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
€ 531.00
Specifications
| Antigen | ARAP1 |
|---|---|
| Clone | ARAP1-2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Biotin |
Description
ARAP1 Monoclonal antibody specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| ARAP1 | |
| Western Blot | |
| Biotin | |
| Mouse | |
| Cell Cycle and Replication | |
| PBS | |
| 116985 | |
| IgG1 | |
| Protein A purified |
| ARAP1-2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2 | |
| His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1 | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title