missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAP1 Antibody (ARAP1-2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68801H
This item is not returnable.
View return policy
Description
ARAP1 Monoclonal antibody specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| ARAP1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunoprecipitation | |
| ARAP1-2 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2 | |
| His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 116985 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction