missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/ARG3.1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 474.00
Specifications
| Antigen | ARC/ARG3.1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677342
|
Novus Biologicals
NBP2-92576-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632402
|
Novus Biologicals
NBP2-92576-0.1ml |
0.1 mL |
€ 474.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARC/ARG3.1 Polyclonal antibody specifically detects ARC/ARG3.1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ARC/ARG3.1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Neuroscience, Sensory Systems | |
| PBS with 50% glycerol, pH7.3. | |
| 23237 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| activity-regulated cytoskeleton-associated protein, ARC/ARG3.1, KIAA0278Arg3.1Activity-regulated gene 3.1 protein homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ARC (NP_056008.1). MELDHRTSGGLHAYPGPRGGQVAKPNVILQIGKCRAEMLEHVRRTHRHLLAEVSKQVERELKGLHRSVGKLESNLDGYVPTSDSQRWKKSIKACLCRCQETIANLERWVKREMHVWREVFYRLERWADRLESTGGKYPVGSESARHTVSVGVGGPESYCHEADGYDYTVSPYAITPPPAAGELPGQEPAEAQQYQPWVPGEDGQPSPGVDTQIFEDPREF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title