missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/NOL3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41753AF594
This item is not returnable.
View return policy
Description
ARC/NOL3 Polyclonal antibody specifically detects ARC/NOL3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ARC/NOL3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ARC. The immunogen is located within the last 50 amino acids of ARC. Amino Acid Squence: PDPEPEPDFEERDESEDS | |
| 0.1 mL | |
| Apoptosis | |
| 8996 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction