Learn More
Arcaine Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Arcaine sulfate |
| CAS | 14923-17-2 |
| Molecular Formula | C6 H16 N6 . H2 O4 S |
| Quantity | 20 mg |
| Synonym | 1,1'-Tetramethylenediguanidine sulfate (1:1), 1,4-Diguanidinobutane sulfate |
| InChI Formula | InChI=1S/C6H16N6.H2O4S/c7-5(8)11-3-1-2-4-12-6(9)10;1-5(2,3)4/h1-4H2,(H4,7,8,11)(H4,9,10,12);(H2,1,2,3,4) |
| SMILES | NC(=N)NCCCCNC(=N)N.OS(=O)(=O)O |
| IUPAC Name | 1-(4-carbamimidamidobutyl)guanidine;sulfuric acid |
| Molecular Weight (g/mol) | 270.31 |
| Formula Weight | 270.111 |