missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARCN1 Monoclonal Antibody (OTI6A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504315S
This item is not returnable.
View return policy
Description
ARCN1 Monoclonal antibody specifically detects ARCN1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| ARCN1 | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48444, Q66H80 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 300674, 372, 479415 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI6A6 | |
| Unconjugated | |
| ARCN1 | |
| 4632432M07Rik; Archain; archain 1; archain vesicle transport protein 1; Arcn1; coatomer delta subunit; coatomer delta-COP; coatomer protein complex, subunit delta; coatomer protein delta-COP; coatomer subunit delta; COPD; delta-coat protein; Delta-COP; nur17; pale coat neuro | |
| Full length human recombinant protein of human ARCN1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction