missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arctiin, TRC
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
Specifications
Specifications
| Chemical Name or Material | Arctiin |
| CAS | 20362-31-6 |
| Molecular Formula | C27 H34 O11 |
| Quantity | 250 mg |
| Synonym | 2(3H)-Furanone, 4-[(3,4-dimethoxyphenyl)methyl]-3-[[4-(β-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-, (3R,4R)-, 2(3H)-Furanone, 4-[(3,4-dimethoxyphenyl)methyl]-3-[[4-(β-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-, (3R-trans)-, Arctiin (7CI,8CI), (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]-3-[[4-(β-D-glucopyranosyloxy)-3-methoxyphenyl]methyl]dihydro-2(3H)-furanone, Arctigenin-4-glucoside, Arctii, NSC 315527 |
| InChI Formula | InChI=1S/C27H34O11/c1-33-18-6-4-14(10-20(18)34-2)8-16-13-36-26(32)17(16)9-15-5-7-19(21(11-15)35-3)37-27-25(31)24(30)23(29)22(12-28)38-27/h4-7,10-11,16-17,22-25,27-31H,8-9,12-13H2,1-3H3/t16-,17+,22+,23+,24-,25+,27+/m0/s1 |
| SMILES | COc1ccc(C[C@H]2COC(=O)[C@@H]2Cc3ccc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(OC)c3)cc1OC |
| IUPAC Name | (3R,4R)-4-[(3,4-dimethoxyphenyl)methyl]-3-[[3-methoxy-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]methyl]oxolan-2-one |
| Molecular Weight (g/mol) | 534.55 |
| Formula Weight | 534.2101 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction