missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody (1A9/5), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08340FR
This item is not returnable.
View return policy
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| ARF1 | |
| Monoclonal | |
| DyLight 680 | |
| ADP-ribosylation factor 1 | |
| C-terminal human ARF1 peptide SNQLRNQ (Uniprot: P84077) | |
| 100 μg | |
| Signal Transduction | |
| 375 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1A9/5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction