missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody (ARFS 1A9/5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 453.00
Specifications
| Antigen | ARF1 |
|---|---|
| Clone | ARFS 1A9/5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18506971
|
Novus Biologicals
NB100-2635 |
0.1 mg |
€ 453.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18523594
|
Novus Biologicals
NB100-2635-0.025mg |
0.025 mg |
€ 190.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| ARF1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS | |
| 375 | |
| IgG2a | |
| Protein A purified |
| ARFS 1A9/5 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADP-ribosylation factor 1 | |
| C-terminal Human ARF1 peptide SNQLRNQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title