missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody (ARFS 1A9/5), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2635JF549
This item is not returnable.
View return policy
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ARF1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ADP-ribosylation factor 1 | |
| C-terminal Human ARF1 peptide SNQLRNQ | |
| 0.1 mL | |
| Signal Transduction | |
| 375 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunofluorescence | |
| ARFS 1A9/5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction