missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody (ARFS 3F1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-85530AF532
This item is not returnable.
View return policy
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
| ARF1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ARF1 | |
| C-terminal Human ARF1 peptide SNQLRNQ | |
| 0.1 mL | |
| 375.0 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| ARFS 3F1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ADP-ribosylation factor 1 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| ARFS 3F1 is Class 1 ARF specific. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction