missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 195.00 - € 322.00
Specifications
| Antigen | ARF3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635186
|
Novus Biologicals
NBP3-12541-100ul |
100 μg |
€ 322.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668108
|
Novus Biologicals
NBP3-12541-50ul |
50 μg |
€ 195.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF3 Polyclonal antibody specifically detects ARF3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| ARF3 | |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 377 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADP-ribosylation factor 3, small GTP binding protein | |
| Produced in rabbits immunized with purified, recombinant Human ARF3 (Uniprot#: P61204; Met1-Lys181). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title