Learn More
ARF4 Polyclonal Antibody
GREENER_CHOICE
ARF4 Polyclonal Antibody
Brand:  Invitrogen PA5109405
Product Code. 17187333
  • € 502.00 / 100µL
Description
ARF4 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ARF4
Polyclonal
Unconjugated
ARF4
AA407803; ADP ribosylation factor 4; ADP-ribosylation factor 2; ADP-ribosylation factor 4; ARF2; Arf4
Rabbit
Affinity Chromatography
RUO
11843, 378, 79120
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
1.41 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
P18085, P61750, P61751
ARF4
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human ARF4 (NP_001651.1).
100 μL
Primary
Human, Mouse, Rat