missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arfaptin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557923
This item is not returnable.
View return policy
Description
Immunogen sequence: ITSHGFDNTK EGVIEAGAFQ GSPAPPLPSV MSPSRVAASR LAQQGSDLIV PAGGQRTQTK SGPVILADEI KNPAMEKLEL VRKWSLNTY Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
ARFIP1 gene ontology annotations related to this gene include regulation of protein secretion.
Specifications
| Arfaptin 1 | |
| Polyclonal | |
| Unconjugated | |
| ARFIP1 | |
| 2310030I10Rik; ADP ribosylation factor interacting protein 1; ADP-ribosylation factor interacting protein 1; ADP-ribosylation factor-interacting protein 1; Arfaptin; arfaptin 1; Arfaptin1; arfaptin-1; Arfip1; ARFP1; D3Ertd598e; HSU52521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53367 | |
| ARFIP1 | |
| Recombinant protein corresponding to Human Arfaptin 1. Recombinant protein control fragment (Product #RP-102760). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction