missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Arfaptin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10651234
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10651234 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10651234 Supplier Invitrogen™ Supplier No. 402400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Arfaptin 2 Polyclonal Antibody for Western Blot, ICC/IF

Plays a role in constitutive metalloproteinase (MMP) secretion from the trans Golgi network. May have important functions during vesicle biogenesis at certain cargo subdomains, which could be predominantly utilized by secreted MMPs, such as MMP7 and MMP2 (PubMed:26507660). Participates also in autophagy by regulating the starvation-depdendent trafficking of ATG9A vesicles which deliver the PI4-kinase to the autophagosome initiation site (PubMed:31204568). In addition, plays a role in NF-kappa-B inhibition by interacting with IKBKB and IKBKG (PubMed:26296658). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arfaptin 2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene ARFIP2
Gene Accession No. P53365, Q6AY65, Q8K221
Gene Alias 2310002N04Rik; ADP ribosylation factor interacting protein 2; ADP-ribosylation factor interacting protein 2; ADP-ribosylation factor interacting protein 2 (arfaptin 2); ADP-ribosylation factor-interacting protein 2; Arfaptin 2; arfaptin-2; Arfip2; Partner of RAC1; partner of RAC1 (arfaptin 2); POR; POR1; Protein POR1
Gene Symbols ARFIP2
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the human Arfaptin 2 protein (ADP-ribosylation factor interacting protein 2, AFP2, Partner of Rac1, or POR1).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23647, 293344, 76932
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.