missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37905-20ul
This item is not returnable.
View return policy
Description
SDHC Polyclonal antibody specifically detects SDHC in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SDHC | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Integral membrane protein CII-3, mitochondrial, PGL3, QPs1, QPs-1, SDH3, succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD, succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa, succinate-ubiquinone oxidoreducatase cytochrome B large subunit | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SDHC (NP_002992.1).,, Sequence:, MAALLLRHVGRHCLRAHFSPQLCIRNAVPLGTTAKEEMERFWNKNIGSNRPLSPHITIYSWSLPMAMSICHRGTGIALSAGVSLFGMSALLLPGNFESYL | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6391 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction