missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38388-100ul
This item is not returnable.
View return policy
Description
ARFGAP1 Polyclonal antibody specifically detects ARFGAP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ARFGAP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ARFGAP1 (NP_001268411.1).,, Sequence:, MASPRTRKVLKEVRVQDENNVCFECGAFNPQWVSVTYGIWICLECSGRHRGLGVHLSFVRSVTMDKWKDIELEKMKAGGNAKFREFLESQEDYDPCWSLQEKYNSRAAALFRDKVVALAEGREWSLESSPAQNWTPPQPRTLPSMVHRVSGQPQSVTASSDKAFEDWLNDDLGSYQGAQGNRYVGFGNTPPPQKKEDDFLNNAMSSLYSGWSSFTTGASR | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55738 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction