missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARFGAP1 Monoclonal Antibody (OTI1F6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525466
This item is not returnable.
View return policy
Description
ARFGAP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
ArfGAP1 is a 406 amino acid containing regulatory protein with an Arf-GAP domain known to be involved in the activation of the GTPase activity of ADP ribosylation factor (ARF). ArfGAP1, a GTPase-activating protein (GAP) for the ADP ribosylation factor 1 (ARF1), promotes the hydrolysis of ARF1-bound GTP and is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles thus playing an important role in membrane trafficking and/or vesicle transport. Dissociation of the coat proteins is required for the fusion of these vesicles with target compartments. The activity of this protein is stimulated by phosphoinosides and inhibited by phosphatidylcholine. Two transcript variants encoding different isoforms have been reported. Ubiquitous expression of ArfGAP1 is seen in most tissues.
Specifications
| ARFGAP1 | |
| Monoclonal | |
| 0.63 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N6T3 | |
| ARFGAP1 | |
| Full length human recombinant protein of ARFGAP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1F6 | |
| Unconjugated | |
| ARFGAP1 | |
| ADP ribosylation factor GTPase activating protein 1; ADP-ribosylation factor 1 GTPase activating protein; ADP-ribosylation factor 1 GTPase-activating protein; ADP-ribosylation factor GTPase activating protein 1; ADP-ribosylation factor GTPase-activating protein 1; AI115377; ARF GAP 1; Arf GAP1; ARF1 GAP; ARF1-directed GTPase-activating protein; Arf1gap; ARFGAP1; EGK_02194; GAP protein; HRIHFB2281; RP11-261N11.1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55738 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction