missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARFGAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15711681 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15711681 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15711681 Supplier Invitrogen™ Supplier No. PA553742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGMGLVSRSS VSHSVLSEMQ VIEQETPVSA KSSRSQLDLF DDVGTFASGP PKYKDNPFSL GESFGSRWDT DAAWGMDRVE EKEPEVTISS IRPISERATN RREVESRSSG LESSEARQKF AGAKAISSD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

ARFGAP2 gene ontology annotations related to this gene include GTPase activator activity; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arfgap2
Gene Accession No. Q3MID3, Q8N6H7, Q99K28
Gene Alias 2310032E02Rik; ADP ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase-activating protein 2; ARF GAP 2; ARFGAP2; GTPase-activating protein ZNF289; IRZ; NBLA10535; Zfp289; Zinc finger protein 289; zinc finger protein 289, ID1 regulated; Znf289
Gene Symbols Arfgap2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARFGAP2. Recombinant protein control fragment (Product #RP-91645).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362162, 77038, 84364
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.