missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGEF2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92229-0.02ml
This item is not returnable.
View return policy
Description
ARFGEF2 Polyclonal antibody specifically detects ARFGEF2 in Human samples. It is validated for Western Blot
Specifications
| ARFGEF2 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldinA-inhibited), ARFGEP2, BIG2brefeldin A-inhibited guanine nucleotide-exchange protein 2, Brefeldin A-inhibited GEP 2, dJ1164I10.1, FLJ23723, PVNH2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-370 of human ARFGEF2 (NP_006411.2). VIQAAAVSPKFVRLKHSQAQSKPTTPEKTDLTNGEHARSDSGKVSTENGDAPRERGSSLSGTDDGAQEVVKDILEDVVTSAIKEAAEKHGLTEPERVLGELECQECAIPPGVDENSQTNGIADDRQSLSSADNLESDAQGHQVAARFSHVL | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 10564 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction