Learn More
Invitrogen™ ARFGEF2 Recombinant Rabbit Monoclonal Antibody (24GB6840)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556969
 View more versions of this product
Product Code. 30275363
  • € 500.00 / 100µL
Description
ARFGEF2 Recombinant Monoclonal Antibody for Western Blot
ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ARFGEF2
Recombinant Monoclonal
Unconjugated
ARFGEF2
ADP ribosylation factor guanine nucleotide exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited); AI463430; Arfgef2; ARFGEP2; AW493672; Big2; Brefeldin A-inhibited GEP 2; Brefeldin A-inhibited guanine nucleotide-exchange factor 2; brefeldin A-inhibited guanine nucleotide-exchange protein 2; dJ1164I10.1; E230011G24Rik; PVNH2
Rabbit
Affinity chromatography
RUO
10564, 296380, 99371
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6840
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
A2A5R2, Q7TSU1, Q9Y6D5
ARFGEF2
A synthesized peptide derived from human ARFGEF2 (aa 1501-1514).